missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ Therma Welder Gloves
Click to view available options
Size: 7
Description
- Black water-repellent heat resistant cowhide grain leather glove
- Gunn cut, winged thumb with yoke, crispin type
- 200 mm split leather cuff
- Stitch with Kevlar™ thread
- Its leather is ideal for welders' gloves
- Winged thumb and flexibility of the leather make this glove anatomical, thus enabling precision work
- The grain leather heat resistant offers good heat resistance and insulation
- The Kevlar™ thread assembly ensures sturdy seams with respect to the heat and wear as well as better thermal insulation
- The water repellent treatement allows a use in wet environement
- Split leather cuff protects a large part of the forearm against mechanical hazards
Specifications
Specifications
Certifications/Compliance | EN 420-03 EEC Basic standards, EN 388-03 Mechanical hazards, EN 407-04 Thermical hazards, EN 12477-02 Welding Type B |
Color | Black |
Material | Cowhide Leather |
Size | 7 |
Thickness (Metric) | 0.8 to 1.1 mm |
Length (Metric) | 39 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction