missing translation for 'onlineSavingsMsg'
Learn More

Theophylline-7-acetic acid, 98%, Thermo Scientific Chemicals

Product Code. 15522363
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 652-37-9
Molecular Formula C9H9N4NaO4
Molecular Weight (g/mol) 260.19
MDL Number MFCD00022832
InChI Key MSFVZSOKOXZSME-UHFFFAOYSA-M
Synonym acefylline, theophylline-7-acetic acid, acephylline, theophyllineacetic acid, carboxymethyltheophylline, 7-carboxymethyl theophylline, 7-theophyllinylacetic acid, theophyllin-7-ylacetic acid, 7-theophyllinessigsaeure, 7-theophyllineacetic acid
PubChem CID 69550
IUPAC Name 2-(1,3-dimethyl-2,6-dioxopurin-7-yl)acetic acid
SMILES [Na+].CN1C2=C(N(CC([O-])=O)C=N2)C(=O)N(C)C1=O

Specifications

Melting Point 270.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 100 g
Formula Weight 238.2
Percent Purity 98%
Chemical Name or Material Theophylline-7-acetic acid
compliance-icons
Product Identifier
  • Theophyllin-7-ylacetic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'