Learn More
WTW™ TFK™ 325 Temperature Sensor
Temperature sensors by WTW are used with pH and ORP electrodes that do not have their own temperature sensor
Brand:  WTW™ 102280
Product Code. 10697291
  • 186.00€ / Each
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
pH and ORP measurements must always have a temperature indication. During pH measurement, the indication of a temperature to compensate the temperature effect, and the documentation, with ORP measurement, only for documentation purposes. During ISE measurement it is important that the calibration and the measurement are performed in the same temperature range. Temperature sensors by WTW are used with pH and ORP electrodes that do not have their own temperature sensor. The temperature sensors are equipped with two 4 mm banana plugs.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Temperature Sensor
1 m
1 m
Yes