Learn More
WTW™ TFK™ 325/HC Temperature Sensor
Temperature sensors by WTW are used with pH and ORP electrodes that do not have their own temperature sensor
Brand: WTW™ 109818
This item is not returnable.
View return policy
Description
pH and ORP measurements must always have a temperature indication. During pH measurement, the indication of a temperature to compensate the temperature effect, and the documentation, with ORP measurement, only for documentation purposes. During ISE measurement it is important that the calibration and the measurement are performed in the same temperature range. Temperature sensors by WTW are used with pH and ORP electrodes that do not have their own temperature sensor. The temperature sensors are equipped with two 4 mm banana plugs.Specifications
For pH electrodes | |
1 m | |
Yes |
Temperature Sensor | |
1 m |