missing translation for 'onlineSavingsMsg'
Learn More

TFF1/pS2 Antibody (TFF1/7891), Novus Biologicals™

Mouse Monoclonal Antibody

249.00€ - 494.00€

Specifications

Antigen TFF1/pS2
Clone TFF1/7891
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18650094
View Documents
Novus Biologicals
NBP3-20561-100ug
100 μg
494.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18684914
View Documents
Novus Biologicals
NBP3-20561-20ug
20 μg
249.00€
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TFF1/pS2 Monoclonal antibody specifically detects TFF1/pS2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

TFF1/pS2
0.2 mg/ml
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Breast Cancer, Cancer
10mM PBS with 0.05% BSA
7031
IgG
Protein A or G purified
TFF1/7891
Immunohistochemistry-Paraffin 1-2 μg/mL
Monoclonal
Purified
RUO
Human
BCEIbreast cancer, estrogen-inducible sequence expressed in, Breast cancer estrogen-inducible protein, breast cancer estrogen-inducible sequence, D21S21, gastrointestinal trefoil protein pS2, hP1.A, HPS2, pNR-2, Polypeptide P1.A, Protein pS2, pS2, trefoil factor 1, trefoil factor, BCE1, human pS2 induced by estrogen from human breast cancercell line M10HP1.A
Recombinant fragment (around aa1-84) of human TFF1/pS2 protein (exact sequence is proprietary)
Primary
Store at 4&drg;C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.