missing translation for 'onlineSavingsMsg'
Learn More
Tetramethylurea, 99%, Thermo Scientific Chemicals Product Code.: 11427757

Tetramethylurea, 99%, Thermo Scientific Chemicals

Product Code. 11427757
50 g
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy

Product Code. 11427757

Brand: Thermo Scientific Alfa Aesar A16295.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tetramethylurea is used as a solvent in dyestuff industries, in condensation reaction and intermediates in surfactant. It is utilized for base catalyzed isomerization and alkylation hydrocyanation due to its low permittivity. It reacts with oxalyl chloride to prepare tetramethyl chloroformamidinium chloride, which is used for the conversion of carboxylic acids and dialkyl phosphates to anhydrides and pyrophosphates respectively.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethylurea is used as a solvent in dyestuff industries, in condensation reaction and intermediates in surfactant. It is utilized for base catalyzed isomerization and alkylation hydrocyanation due to its low permittivity. It reacts with oxalyl chloride to prepare tetramethyl chloroformamidinium chloride, which is used for the conversion of carboxylic acids and dialkyl phosphates to anhydrides and pyrophosphates respectively.

Solubility
Miscible with water.Miscible with water, petroleum ether and common solvents.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 632-22-4
Molecular Formula C5H12N2O
Molecular Weight (g/mol) 116.164
MDL Number MFCD00008319
InChI Key AVQQQNCBBIEMEU-UHFFFAOYSA-N
Synonym tetramethylurea, temur, urea, tetramethyl, n,n,n',n'-tetramethylurea, tetramethyluree, tmu, urea, 1,1,3,3-tetramethyl, urea, n,n,n',n'-tetramethyl, tetramethyluree french, tetramethyl-ureaShow MoreShow Less
PubChem CID 12437
ChEBI CHEBI:84278
IUPAC Name 1,1,3,3-tetramethylurea
SMILES CN(C)C(=O)N(C)C

Specifications

Melting Point -1°C
Density 0.969
Boiling Point 176°C to 178°C
Flash Point 75°C (167°F)
Odor Mild
Linear Formula (CH3)2NCON(CH3)2
Refractive Index 1.4506
Quantity 50 g
Beilstein 773898
Merck Index 14,9230
Solubility Information Miscible with water;Miscible with water,petroleum ether and common solvents.
Formula Weight 116.16
Percent Purity 99%
Chemical Name or Material Tetramethylurea
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10242973 Certificate TypeCertificate of Analysis Date22/08/2022 Catalog NumberA16295.18,A16295.30
Lot Number10152115 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16295.18,A16295.30
Lot NumberH31Y071 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16295.18,A16295.30
Lot Number10222154 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16295.18,A16295.30
Lot NumberF05Z027 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16295.18,A16295.30
compliance-icons compliance-icons
Product Identifier
  • Tetramethylurea
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1B
Hazard Statement
  • H302-Harmful if swallowed.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Tetramethylurea

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetramethylurea, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.