missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofurfuryl bromide, 95%, Thermo Scientific Chemicals

Product Code. 11462208
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1192-30-9
Molecular Formula C5H9BrO
Molecular Weight (g/mol) 165.03
MDL Number MFCD00016894
InChI Key VOHILFSOWRNVJJ-UHFFFAOYNA-N
Synonym tetrahydrofurfuryl bromide, 2-bromomethyl tetrahydrofuran, 2-bromomethyl oxolane, furan, 2-bromomethyl tetrahydro, 2-bromomethyl-tetrahydrofuran, furan, tetrahydro-2-bromomethyl, tetrahydrofurfurylbromide, tetrahydrofrufryl bromide, 2-bromomethyltetrahydrofuran
PubChem CID 14501
IUPAC Name 2-(bromomethyl)oxolane
SMILES BrCC1CCCO1

Specifications

Density 1.452
Boiling Point 169°C to 171°C
Quantity 250 g
Refractive Index 1.488
Beilstein 102717
Formula Weight 165.04
Percent Purity 95%
Chemical Name or Material Tetrahydrofurfuryl bromide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'