Learn More
WTW™ TetraCon™ Graphite 4-Electrode Conductivity Cell
WTW carries two special measuring cells, which are specifically geared for these applications: The TetraCon™ S consists of an electrode body, which is perfectly fitted to measure pasteous samples thanks to its shovel shape and is easy to clean. The TetraCon™ 325/C is fitted with a chemical-resistant head made of PEEK to measure especially acidic samples.
Brand:  WTW™ 301602
Product Code. 10159271
  • 754.00€ / Each
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
Specifications
4-electrode-conductivity cell
Eliminate errors caused by polarization effects, and from dirty samples
Graphite
1.5 m
Special TetraCon™ 325/S Electrode System
Built-in temperature sensor, 4.9 ft. cable, waterproof plug
0°C to 100°C
15.3 mm