Learn More
WTW™ TetraCon™ DU/T Flow-through Conductivity Cell
Flow measuring cells by WTW for medium and high conductivity are suitable for extensive measuring series in the lab but also for the maintenance and measurement of cleaning solutions in special systems.
Brand:  WTW™ 301252
Product Code. 10011941
  • 945.00€ / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
0.778 cm-1
Epoxy, Graphite
TetraCon™ DU/T
Flow-through Conductivity Cell
4-electrode, temperature sensor
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products