Learn More
WTW™ TetraCon™ 925-3 4 Electrode Conductivity Cell
With its easy handling and robust design, these electrode conductivity cells are proven 2 and 4 electrode technology.
Brand:  WTW™ 301711
Product Code. 11725184
  • 677.00€ / Each
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • IDS conductivity measuring cells – measurements from pure water all the way to concentrated solutions with proven two and four electrode technology
  • Proven 2 and 4 electrode technology
  • Easiest handling, robust design
  • Wide area of application - from pure water to highly concentrated solutions
TRUSTED_SUSTAINABILITY
Specifications
Specifications
4-electrodes, Graphite
0°C to 100°C
120 mm
3 m
15.3 mm