Learn More
Testo™ Testoterm™ Self-Adhesive Thermometer Strips
Thin, temperature-sensitive pieces of film that change color when the temperature exceeds a specific value. This makes them ideal for monitoring the temperature of products and processes where the temperature should not exceed a certain limit.
Brand: Testo™ 0646 3341
This item is not returnable.
View return policy
Description
- The temperature strips are available in booklets of 10. The strips are peeled off and applied to the object you wish to measure
- The strips have a temperature scale ranging from +204 °C to +260 °C which is divided into little white boxes
- As soon as a specific temperature has been exceeded, the strip changes color in the relevant boxes in the space of 2 to 3 seconds
- The temperature steps for the individual boxes are as follows: 204 °C, 210 °C, 216 °C, 224 °C, 232 °C, 241 °C, 249 °C, 254 °C and 260 °C.
- When the strips change color, they do so irreversibly. When a certain temperature has been exceeded, the color does not change back again when the temperature returns to previous levels
- This allows you to see that critical thresholds have been exceeded even when the temperatures have returned to the levels they were before the limit was reached. This makes them ideal for use in storage and transport.
Specifications
50 x 18 mm | |
204°C to 260°C | |
±1% of mv ±1°C |