Learn More
Elma™ Test Tube Holders
Ensures that tubes can be securely positioned and arranged in the Elma Basket.
65.00€ - 103.00€
Specifications
Type Test Tube Holder
For Use With (Equipment) Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY
Material Stainless Steel
View More Specs
Products 3
Product Code Brand Model Price Quantity & Availability  
Product Code Brand Model Price Quantity & Availability  
17789899
View Documents
Elma™
1111973
30/80
65.00€
Each
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
17851297
View Documents
Elma™
1112084
100/120
88.15€
Each
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
17801357
View Documents
Elma™
1112123
150/300
103.00€
Each
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
Description
Description
  • Diverse Elma holders ensure that tubes can be securely positioned and arranged in the Elma Basket
  • Optimum positioning of the items inside the bath is also a prerequisite for achieving the best possible cleaning results
  • Allows items to be individually positioned
  • Lets you clearly structure your cleaning process
  • Prevents scratches on the tank floor
  • Holders are available for various applications
  • Special solutions are available (e.g. for conical flasks)
Specifications
Specifications
Test Tube Holder
Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY
Stainless Steel