missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Semi-Quantitative Test Strips, QUANTOFIX Muntistick

Semi-quantitative test strips for the determination of total hardness, carbonate hardness, and pH. Ideal for rapid and easy analysis of aquarium water quality.

Brand:  Macherey-Nagel™ 91327

Product Code. 10267890

  • 24.40€ / Pack of 25

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Carbonate hardness gradation: 0, 3.8, 7.5, 12.5, 18.8, 25.0 °e
  • pH gradation: 6.4, 6.8, 7.2, 7.6, 8.0, 8.4
  • Total hardness gradation: 0, 6.3, 12.5, 18.8, 25.0, 31.3 °e
  • Carbonate hardness color change: bright green → blue
  • pH color change: yellow → red
  • Total hardness color change: green → red
Specifications

Specifications

Carbonate hardness, pH, Total hardness
25 test strips in a tube, instruction leaflet
25 Tests
4°C to 30°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions