Learn More
Mecmesin™ Test Stand for 59885-Series Force Gauge
Accessory for 59885-Series force gauge.
Brand:  Mecmesin™ 804001
Encompass
Product Code. 11907979
Description
Description
  • Repeatable test results
  • Single-column motorized test stands
  • Crosshead speed: 1 to 1000mm/min
  • Single cycle test mode with full-speed crosshead return
  • Auto-travel reverse on alarm point or sample break
Specifications
Specifications
Test Stand
Accessory for 59885-Series Force Gauge
Measuring force generated during compression
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products