Learn More
WTW™ Test Agent for OxiTop™ Systems
For use with Monitoring OxiTop™ systems; these test agents can be used for checking 6 measuring systems.
Brand:  WTW™ 209333
79.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 12333790
  • 83.00€ / Pack of 8
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
    • Includes calibration tablets
    • For checking 6 measuring systems (long term test of leakage testing and negative pressure)
    • For use with Monitoring OxiTop™ systems
    • Test agent product type
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Test Agent
For checking 6 measuring systems (long term test of leakage testing and negative pressure)
Monitoring OxiTop™ systems
Calibration tablets
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
WTW™ Test Agent for OxiTop™ Systems > Includes: Calibration tablets; For use with: Checking 6 measuring systems