missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 mL
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed in cross couplig reactions.
Solubility
Reacts with water.
Notes
Handle under dry protective gas. Air sensitive. Store away from strong bases and oxidizing agents, heat, acids and wate
It is employed in cross couplig reactions.
Solubility
Reacts with water.
Notes
Handle under dry protective gas. Air sensitive. Store away from strong bases and oxidizing agents, heat, acids and wate
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Spezifikation
CAS | 7565-59-5 |
Density | 0.966 |
Molecular Formula | C4H9BrZn |
Linear Formula | (CH3)3CZnBr |
MDL Number | MFCD00671989 |
Quantity | 50 mL |
UN Number | UN3399 |
Sensitivity | Air sensitive; May form a precipitate |
Synonym | tert-butylzinc bromide, bromo-t-butylzinc, bromo tert-butyl zinc, zinc, bromo 1,1-dimethylethyl, tert-butylzinc bromide, 0.50 m in thf, tert-butylzinc bromide 0.5 m in tetrahydrofuran, tert-butylzinc bromide solution, 0.5 m in thf, tert-butylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles |
Solubility Information | Reacts with water. |
Mehr anzeigen |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'