missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-butyl (S)-4-(dimethylamino)-1-(piperazine-1-carbonyl)butylcarbamate, Thermo Scientific™
46.40€ - 625.00€
Chemical Identifiers
CAS | 1174064-68-6 |
---|---|
Molecular Formula | C16H32N4O3 |
Molecular Weight (g/mol) | 328.457 |
InChI Key | CDZDEHVWAXDQAQ-ZDUSSCGKSA-N |
Synonym | tert-butyl s-4-dimethylamino-1-piperazine-1-carbonyl butylcarbamate, tert-butyl n-2s-5-dimethylamino-1-oxo-1-piperazin-1-yl pentan-2-yl carbamate, tert-butyl 2s-5-dimethylamino-1-oxo-1-piperazin-1-yl pentan-2-yl carbamate |
PubChem CID | 45594327 |
IUPAC Name | tert-butyl N-[(2S)-5-(dimethylamino)-1-oxo-1-piperazin-1-ylpentan-2-yl]carbamate |
SMILES | CC(C)(C)OC(=O)NC(CCCN(C)C)C(=O)N1CCNCC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10587770
|
Thermo Scientific Maybridge
MO08535CB |
250 mg | Amber Glass Bottle |
46.40€
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10301380
|
Thermo Scientific Maybridge
MO08535DA |
1 g | Amber Glass Bottle |
153.00€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10687390
|
Thermo Scientific Maybridge
MO08535DE |
5 g | Amber Glass Bottle |
625.00€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
1174064-68-6 | |
328.457 | |
tert-butyl s-4-dimethylamino-1-piperazine-1-carbonyl butylcarbamate, tert-butyl n-2s-5-dimethylamino-1-oxo-1-piperazin-1-yl pentan-2-yl carbamate, tert-butyl 2s-5-dimethylamino-1-oxo-1-piperazin-1-yl pentan-2-yl carbamate | |
tert-butyl N-[(2S)-5-(dimethylamino)-1-oxo-1-piperazin-1-ylpentan-2-yl]carbamate |
C16H32N4O3 | |
CDZDEHVWAXDQAQ-ZDUSSCGKSA-N | |
45594327 | |
CC(C)(C)OC(=O)NC(CCCN(C)C)C(=O)N1CCNCC1 |
Specifications
1174064-68-6 | |
C16H32N4O3 | |
CDZDEHVWAXDQAQ-ZDUSSCGKSA-N | |
tert-butyl N-[(2S)-5-(dimethylamino)-1-oxo-1-piperazin-1-ylpentan-2-yl]carbamate | |
45594327 | |
97% |
97% | |
tert-butyl s-4-dimethylamino-1-piperazine-1-carbonyl butylcarbamate, tert-butyl n-2s-5-dimethylamino-1-oxo-1-piperazin-1-yl pentan-2-yl carbamate, tert-butyl 2s-5-dimethylamino-1-oxo-1-piperazin-1-yl pentan-2-yl carbamate | |
CC(C)(C)OC(=O)NC(CCCN(C)C)C(=O)N1CCNCC1 | |
328.457 | |
328.45 | |
tert-Butyl (S)-4-(dimethylamino)-1-(piperazine-1-carbonyl)butylcarbamate |
Safety and Handling
Danger