Learn More
3M™ Nomad™ Terra Heavy Traffic Entrance Matting 8100
Entrance coiled vinyl scraper matting designed for heavy traffic applications. The construction allows dirt and moisture to flow through the matting so it is not retracked into the building. Grease and oil resistant.
Brand:  3M™ Nomad™ 7100116706
 View more versions of this product
Product Code. 17118730
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • Unbacked scraper matting designed for high traffic applications
  • High quality vinyl offers excellent crush resistance
  • Unbacked, open construction of the mat allows water to drain and soil to pass through, out of sight
  • Easily vacuum out trapped dirt
TRUSTED_SUSTAINABILITY
Specifications
Specifications
610 x 122 cm
Nomad™ Terra Dirt Control Mats
Brown
Elastic Vinyl
Heavy traffic areas
17 mm
610 cm
122 cm