missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Teriflunomide
GREENER_CHOICE

Product Code. 18780298
Click to view available options
:
50mg
This item is not returnable. View return policy

Product Code. 18780298

Brand: Tocris Bioscience™ 5069/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dihydroorotate dehydrogenase inhibitor

Teriflunomide is an inhibitor of dihydroorotate dehydrogenase (Kd = 12 nM; Ki = 179 nM). Inhibits proliferation of mitogen- or cytokine-stimulated lymphoid cells in vitro by inhibiting cell cycle progression from G1 to S. Immunosuppressive agent. Active metabolite of leflunomide.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 mg
Product Type Teriflunomide
Purity >99%
Molecular Weight (g/mol) 270.21
Inhibitors Teriflunomide

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.