missing translation for 'onlineSavingsMsg'
Learn More

Terbium powder, -40 mesh, 99.9% (REO)

Product Code. 15480957
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 15480957

Brand: Thermo Scientific Alfa Aesar 000322.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 7440-27-9 | Tb | 158.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged in air stable poly bottle
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-27-9
Molecular Formula Tb
Molecular Weight (g/mol) 158.93
MDL Number MFCD00011256
InChI Key GZCRRIHWUXGPOV-UHFFFAOYSA-N
Synonym unii-06ssf7p179, ingot, foil, terbio, atom, chips, pieces, powder, acmc-20ajly, pieces, distilled dendritic
PubChem CID 23958
ChEBI CHEBI:33376
IUPAC Name terbium
SMILES [Tb]

Specifications

Color Silver to Gray
Quantity 5 g
Mesh Size −40 Mesh
Odor Odorless
Packaging Poly bottle
Sensitivity Air and moisture sensitive
Form Powder
UN Number UN3089
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Terbium powder
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.