Learn More
Testo™ Testoterm™ Temperature Strips
Use these strips for monitoring the temperature of moving or small objects and long-term monitoring of transport and storage temperatures. Testoterm™ Temperature Strips can be applied to a variety of different surfaces and react to specific increases in temperature by changing color irreversibly within 2 to 3 seconds.
Brand:  Testo™ 0646 0916
 View more versions of this product
Product Code. 10282161
  • 40.45€ / Pack of 10
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Self-adhesive temperature strips
  • Available in booklets of 10
  • Strips are peeled off and applied to the object you wish to measure
  • Change color when thresholds are exceeded within 2 to 3 seconds
Specifications
Specifications
Self-adhesive
±1.5°C
71°C to 110°C
Self-adhesive Temperature Strips