Learn More
Aalborg™ Teflon Standard Valve
Aalborg™ Needle Valve for T-Style Flowmeter is designed for laboratory and industrial applications for regulating corrosive gases and liquids or for high purity service.
Brand: Aalborg™ VCLTT
This item is not returnable.
View return policy
Description
- Fluids in contact with PTFE only
- Structurally rigid metal shell.
- One PTFE o-ring
- Simplicity - only seven components.
Specifications
2400 mL/min. | |
65°C | |
130mL/min. | |
Standard Valve |