Learn More
Aalborg™ Teflon Standard Valve
Aalborg™ Needle Valve for T-Style Flowmeter is designed for laboratory and industrial applications for regulating corrosive gases and liquids or for high purity service.
Brand:  Aalborg™ VCLTT
Product Code. 11768378
  • 56.00€ / Each
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Description
  • Fluids in contact with PTFE only
  • Structurally rigid metal shell.
  • One PTFE o-ring
  • Simplicity - only seven components.
Specifications
Specifications
2400 mL/min.
65°C
130mL/min.
Standard Valve