Learn More
Bochem Teclu burner f. Natural gas, w., Stopcock, 1300 °C
Brand:  Bochem 7150
Teclu burner f. Natural gas, w., Stopcock, 1300 °
Product Code. 10004930
  • 187.00€ / Each
Estimated Shipment: 03-04-2025