missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Technical Calibration Buffer pH 6.86, Hanna Instruments™
Click to view available options
Quantity:
20 mL
Specifications
Specifications
Color | Colorless |
pH | 6.86 |
Physical Form | Liquid |
Quantity | 20 mL |
Solution Type | Buffer |
Formulation | Aqueous Buffer Solution |
Odor | Odorless |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction