missing translation for 'onlineSavingsMsg'
Learn More

Scotsman™ TCS 180 AS Cublet Ice Maker and Dispenser

TCS 180 AS cublet ice maker easily dispenses both ice and water.

Brand:  Scotsman™ TCS180AS

Product Code. 11734788

  • 5870.00€ / Each
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket

Récupel-bijdrage uitgesloten
This item is not returnable. View return policy

Description

Description

TCS 180 AS cublet ice maker also dispenses both ice and water.

  • Allow customers to cool their drinks as per their own taste, making it easy and preventing any potential sanitary hazards
  • Clear and pure ice bits drop directly into the consumer's glass, with no manipulation
  • Industry benchmark in the ice or ice and water dispensers segment
  • Holds 5 g of 'Cubelet' super compressed ice
Specifications

Specifications

5 g
600 mm
34.4 in.
50 Hz
230 V
134 lb.
15.4 in.
23.6 in.
Ice Making and Dispensing
874 mm
1 to 5 bar
2-year
61 kg
390 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.