missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Tat-Beclin 1 L11 Autophagy Inducing Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
1 mg
5 mg
Unit Size:
0.1mg
1mg
5mg
Specifications
Specifications
| Species | Non-species Specific |
| Content And Storage | Store at -20°C in powder form. Store at -80°C once reconstituted. |
| Gene Alias | Autophagy Inducing peptide, Tat-Beclin, Tat-Beclin 1, Tat-Beclin 1 peptide, Tat-Beclin peptide, Tat-L11 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 3.08 kDa |
| Quantity | 0.1 mg |
| Research Category | Autophagy |
| Storage Buffer | This product is supplied lyophilized. Purity is ≥ to 97% (HPLC) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction