missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tango™ CXCR1-bla U2OS Cells

Product Code. 10135223
Click to view available options
Quantity:
8 x 106 Cells
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10135223

Brand: Thermo Scientific™ K1807

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

The Tango™ CXCR1-bla U2OS cells contain the human Chemokine (C-X-C motif) receptor 1 (CXCR1) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line.

The Tango™ CXCR1-bla U2OS cells contain the human Chemokine (C-X-C motif) receptor 1 (CXCR1) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase (bla) reporter gene under the control of a UAS response element.

The Tango™ CXCR1-bla U2OS cells are functionally validated for Z' and EC50 concentrations of IL-8. In addition, Tango™ CXCR1-bla U2OS cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.

For Academic pricing please login or contact us at discoverysciences@invitrogen.com.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_000634.2
Assay Entry Cell-based beta-arrestin recruitment
Cell State Dividing Cells
Content And Storage LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.

Includes:
• Tango™ CXCR1-bla U2OS cells
• Tango™ CXCR1-bla U2OS Assay Protocol
• Certificate of Analysis

The Tango™ CXCR1-bla U2OS cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Cytoplasmic Tail Native Tail
Description Tango™ CXCR1-bla U2OS Cells, 8 x 106 Cells, U2OS Cell Line, Fluorescent Detection Method, GPCRs Druggable Target, BLA (Beta-Lactamase) Reporter, Liquid Nitrogen Storage
Detection Method Fluorescence
For Use With (Application) FRET
Gene Symbol CXCR1
Includes Tango™ CXCR1-bla U2OS cells (LiveBLAzer™-FRET B/G loading kit, solution D and other materials are required separately; please refer to the protocol). Includes: Tango™ CXCR1-bla U2OS cells, Tango™ CXCR1-bla U2OS assay protocol, certificate of analysis
Population Status Pool of Stable Cells
Product Line GeneBLAzer, Tango
Quantity 8 x 106 Cells
Readout End Point
Reporter Gene BLA (Beta-Lactamase)
Sample Type U2OS
Shipping Condition Room Temperature
Target Entry CXCR1
Type U2OS Cell
Cell Line U2OS
System Type Tango™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.