missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Köttermann Tall Acid and Base Cabinet
Constructed with durable polypropylene, this Tall Acid and Base Cabinet is built to endure the aggressive conditions produced by corrosive acid fumes. Liquid-tight pull-out trays offer easy access while minimizing health and safety risks.
2973.30€ - 4500.00€
Specifications
Color | White |
---|---|
Dimensions (D x W x H) | 515 x 600 x 1900 mm |
Material | Polypropylene |
Door Style | Hinged, Drawer |
For Use With (Application) | Acid and Base Storage |
Product Code | Brand | Configuration | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Configuration | Includes | Price | Quantity & Availability | ||||
17392149
|
Köttermann
377-0001 |
2 Left-hinged Doors, 4 Drawers | No Ventilation Fan | N/A | |||||
17312159
|
Köttermann
377-0003 |
2 Left-hinged Doors, 4 Drawers | Ventilation Fan | N/A | |||||
17302159
|
Köttermann
377-0002 |
2 Right-hinged Doors, 4 Drawers | No Ventilation Fan | N/A | |||||
17322159
|
Köttermann
377-0004 |
2 Right-hinged Doors, 4 Drawers | Ventilation Fan |
4500.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Safe storage of corrosive non-flammable hazardous substances, with acids and bases separated
- Cabinet carcasses and pull-out trays made of resistant polypropylene
- Liquid-tight drawers on low-friction PTFE strips with pull-out stop
- Removal of aggressive vapors via optional ventilation with an external fan
- Corrosion-resistant locks and hinges
- Ideal for installation under high-performance fume cupboards
Specifications
White | |
Polypropylene | |
Acid and Base Storage |
515 x 600 x 1900 mm | |
Hinged, Drawer |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title