Learn More
Sartorius Tacta™ Mechanical Pipettes
Shop All Sartorius Products
Click to view available options
Number of Channels:
1
8
12
Volume (Metric):
0.1 to 3 μL
0.5 to 10 μL
10 to 100 μL
100 to 1000 μL
1000 to 10,000 μL
2 to 20 μL
20 to 200 μL
30 to 300 μL
5 to 100 μL
500 to 5000 μL
Color:
Blue
Gray
Green
Orange
Red
Yellow
Pakningsstørrelse:
Each
Beskrivelse
- Comfortable to hold due to the ergonomically designed handle
- Unique Sartorius Optilock system provides flexibility for volume adjustment and locking
- Controlled and smooth tip ejection with the new Sartorius Optiject technology
- Calibration adjustment to provide accurate results for various liquid types
- Easy to clean, with only three parts to disassemble
- Reliable results, even over lengthy pipetting periods
Tekniske data
Tekniske data
| Compatible Tips | Optifit 10 μL, SafetySpace Filter Tips 10 μL |
| Adjustable Tip Spacing | Yes |
| Autoclavable | Autoclavable |
| Color | Gray |
| Disposable | Reusable |
| Grip Type | Ergonomic |
| Material | Polypropylene |
| Number of Channels | 1 |
| Product Line | Tacta |
| Programmability | No |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion