Learn More
SI Analytics™ TA 20 Plus Interchangable Unit
SI Analytics™ TA 20 Plus Interchangeable Unit is a brown glass bottle with a UV-protection coating for titrants.
Brand:  SI Analytics™ 285211054
Product Code. 10103212
  • 954.00€ / Each
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Features a brown UV-protection coat
  • Brown glass bottle for titrant
  • GL 45 thread size
  • Includes S 40 bottle adapter, hoses, drip tube, and titration tip
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Exchange Unit
Reagent bottle
20 mL