Learn More
Copan Italia™ T-Shaped Plastic Spreaders
Sterile, T-shaped spreaders for dispersing liquids across the surface of agar culture plates. Copan Italia™ T-Shaped Plastic Spreaders feature smooth, rounded surfaces to prevent agar cutting or gouging.
Brand:  Copan Italia™ 173CS05
Product Code. 11756548
  • 198.00€ / Pack of 1000
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Used for performing bacterial CFU plate counts on water, milk, urine, and other liquid samples.
  • Smooth, rounded surfaces prevent cutting or gouging of agar during inoculation of culture plates.
  • T-shape design facilitates even pressure application on the agar surface.
Specifications
Specifications
Single-use
5/Peel pack, 1000/Cs
T
Spreader
Used for performing bacterial CFU plate counts on water, milk, urine and other liquid samples.
171 mm
1000/Pk.
Plastic
Sterile