Learn More
IKA T 25 Digital Ultra-Turrax™ Immersion Blender
GREENER_CHOICE
High-performance immersion blender. IKA™ T 25 Digital Ultra-Turrax™ offers a wide speed range that enables users to work at high circumferential speeds even with small rotor diameters.
Brand:  IKA 0003725000
Product Code. 13326309
  • 2612.00€ / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Récupel-bijdrage uitgesloten
Description
Description
  • Digital speed display
  • Electronic speed control
  • Electronic overload protection
  • Stainless steel dispersing elements can be cleaned quickly and easily
  • Plastic disposable dispersing elements are available in two sizes
  • Error code display
  • Quiet operation
  • EU plug
Specifications
Specifications
3000 to 25,000 rpm
230 V
1 mL to 2 L
EU Plug
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products