missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SZL P1-41
GREENER_CHOICE

Product Code. 18794828
Click to view available options
:
50mg
This item is not returnable. View return policy

Product Code. 18794828

Brand: Tocris Bioscience™ 5076/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective Skp2 inhibitor; suppresses E3 ligase activity

SZL P1-41 is a Skp2 inhibitor; thought to prevent assembly of Skp2-Skp1 complexes. Selectively suppresses Skp2 SCF E3 ligase activity but exhibits no effect on the activity of other SCF complexes. Also inhibits Skp2-mediated p27 and Akt ubiquitination in vivo and in vitro. Suppresses the survival of cancer cells and cancer stem cells by triggering cell senescence and inhibiting glycolysis. Exhibits antitumor effects in multiple animal models and cancer cell lines.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 mg
Product Type SZL P1-41
Purity >98%
Molecular Weight (g/mol) 420.52
Inhibitors SZL P1-41

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.