Learn More
Waters Corporation Symmetry Shield RP18 Column
Reversed-phase columns contain patented embedded polar group technology that shields the silica residual silanol surface from highly basic analytes.
Brand:  Waters Corporation 186000109
Encompass
 View more versions of this product
Product Code. 10753933
  • 1297.50€ / Each
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Description
  • The reduced silanol activity for the SymmetryShield RP18 column significantly improves peak shape and resolution.
Specifications
Specifications
4.6 mm
6000 psi (415 Bar)
5 μm
1/Ea.
Yes
150 mm
Spherical
100 Å
L1
Column