Learn More
Novus Biologicals™ Surfactant Protein A Antibody Pair
Brand: Novus Biologicals™ H00653509-AP42
864.00 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
3 ng/mL to 100 ng/mL | |
Sandwich ELISA | |
653509 | |
1 Set | |
COLEC4, PSAP, PSPA, PSP-A, SFTP1, SFTPA1B, SPA, SP-A, SPA1, SP-A1, surfactant protein A1B | |
1 x PBS, pH 7.4 |
Unconjugated | |
Sandwich ELISA | |
SFTPA1 | |
Aliquot and store at -20°C to +80°C. Avoid freeze/thaw cycles. | |
Human |