Learn More
Thermo Scientific™ SureTect™ Real-Time PCR System
Product Code. p-6017006
Click to view available options
Description:
PikoReal 24 (24-well Piko PCR Plate)
For Use With (Application):
SureTect Real Time PCR Assays
Unit Size:
Each
Product Code. 15234101
Brand: Thermo Scientific™ PT0500
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Quickly and accurately detect microorganisms in a broad range of food and environmental samples using the Thermo Scientific™ SureTect™ System.
Quickly and accurately detect microorganisms in a broad range of food and environmental samples using the Thermo Scientific™ SureTect™ System. The system combines a real-time PCR instrument with bespoke software and convenient pre-filled reagent components that, together with a single PCR protocol for a range of common targets and sample types, streamline the test workflow.
The SureTect System is a unique solution combining speed and performance in an easy-to-use, cost-effective platform—giving you results you can trust.
  • Fast Time to Result: Takes less than 24 hours from sample enrichment to results
  • Single Enrichment Step: Requires no secondary enrichment or regrowth, for faster, simpler testing
  • Easy-to-Use Assay: Requires few pipetting steps, pre-filled lysis tubes and freeze-dried PCR pellets, with no centrifuge or vortex required for sample preparation
  • Streamlined Protocol: Facilitates processing of multiple assays in the same run, for maximum efficiency
  • Superior PCR Technology: Uses probe-based real-time assays for unparalleled sensitivity and specificity
  • Simple, Intuitive Software: Enables rapid, straightforward PCR set-up, automatic data interpretation and traceability of results
  • Small Instrument Footprint: Fits easily into laboratory environments, where bench space is at a premium
TRUSTED_SUSTAINABILITY
Specifications
Thermal Accuracy ±2°C
Sensitivity 1 Copy (Theoretical)
Dynamic Range 10 logs
Content And Storage PikoReal 24
Height (Metric) 310 mm
Width (Metric) 300 mm
Depth (Metric) 230 mm
Thermal Range 4° to 99.9°C
For Use With (Application) SureTect Real Time PCR Assays
Description PikoReal 24 (24-well Piko PCR Plate)
Max. Heating Rate (Thermal Block) >5°C/s
Scan Time for Four Multiplexing Channels <10 s.
Temperature Range (Heated Lid) 30 to 110°C
Max. Cooling Rate 4.5deg;C/s
Excitation 5 LEDs
Detection Range 520 to 740 nm
Software Analysis Modes Absolute quantification; Relative quantification; Melt curve analysis; Allelic discrimination; High Resolution Melt (HRM)
Control (Heated Lid) Automatic Temperature and Pressure Setting
Volume (Metric) Thermal Block Sample 10 to 50μL
Multiplex Up to 4 Targets
Weight 10 kg
Block Configurations 24-well