missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ SureOne™ Aerosol Barrier Pipette Tips
GREENER_CHOICE

Product Code. 13448236 Shop All Fisherbrand Products
Click to view available options
Volume (Metric):
0.1 to 10 μL
0.1 to 20 μL
10 to 100 μL
100 to 1000 μL
2 to 20 μL
20 to 200 μL
30 to 300 μL
Tip Style:
Beveled
Extended Length, Micro Point, Low Retention
Micro Point
Micro Point, Extended Length
Color:
Clear
Natural
Unit Size:
Pack of 4800
Pack of 960
This item is not returnable. View return policy

Product Code. 13448236

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Choose from a comprehensive line of pipet tips for demanding liquid-handling applications

Universal fit for research grade pipetters.

  • Engineered for increased accuracy and are compatible with single-channel, multichannel, and electronic pipetters
  • DNase-/RNase-free; sterile tips are also pyrogen-free
  • Sterile and clear color
  • Filter eliminates aerosol cross-contamination without sealing or trapping samples
  • Tips allow for noncontaminated sample recovery
  • Not autoclavable

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 20 to 200 μL
Filtered Filtered
Sterility Sterile
Color Natural
Autoclavable Not Autoclavable
Length (English) 2 in.
Length (Metric) 58 mm
Format Hinged Rack
Tip Style Micro Point
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line SureOne
Graduations 10 μL, 50 μL, 100 μL
Show More Show Less

As part of our commitment to continuous product improvement, your delivery may contain products with some minor color variation, but their form, fit and function will not be affected. Any questions contact your sales representative.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.