Learn More
Fisherbrand™ SureOne™ Aerosol Barrier Pipette Tips
GREENER_CHOICE
Choose from a comprehensive line of pipet tips for demanding liquid-handling applications
Brand:  Fisherbrand™ TF113-20-FIS
 View more versions of this product
Product Code. 13428236
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
Choose from a comprehensive line of pipet tips for demanding liquid-handling applications
Universal fit for research grade pipetters.
  • Engineered for increased accuracy and are compatible with single-channel, multichannel, and electronic pipetters
  • DNase-/RNase-free; sterile tips are also pyrogen-free
  • Sterile and clear color
  • Filter eliminates aerosol cross-contamination without sealing or trapping samples
  • Tips allow for noncontaminated sample recovery
  • Not autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2 to 20 μL
Sterile
Not Autoclavable
46 mm
Beveled
SureOne
Filtered
Natural
2 in.
Hinged Rack
Universal Fit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ SureOne™ Aerosol Barrier Pipette Tips > 2 to 20μL; Beveled tip