Learn More
Thermo Scientific™ Super Mega Stainless Steel Molds
Product Code. p-4524723 Shop All Epredia Products
Click to view available options
Depth (Metric):
15 mm
Unit Size:
Pack of 5
Product Code. 12618396
Brand: Thermo Scientific™ E10/M8
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Easily accomodate large specimen sizes with Thermo Scientific™ Super Mega Stainless Steel Molds.
  • Sizes fit most cassettes
  • Available in packs of 5
  • Rounded corners
TRUSTED_SUSTAINABILITY
Specifications
Product Type Base Mold
Material Stainless Steel
Length (Metric) 60 mm
Width (Metric) 45 mm
Depth (Metric) 15 mm