Learn More
Sulfuric Acid, 0.005M, Chem-Lab
Product Code. 11973283
Click to view available options
Quantity:
5 L
25 L
For laboratory use, Titrations and TMAR Sulfates determination, ASTM D-3228
Chemical Identifiers
CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O
Specifications
Concentration 0.491g H2SO4 / 1L H2O
Density 1.00g/mL
Packaging PE
Normality 0.01N (±0.00002 at 20°C)
Quantity 25 L
Formula Weight 98.08g/mol
Chemical Name or Material Sulfuric acid