missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfate Standard Solution, 1000μg/mL, Ion HIQU, Chem-Lab
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Description
This sulfate standard solution contains a precise concentration of 1000μg/mL SO42− (±7μg at 20°C). It is used to verify the accuracy of sulfate determinations and to calibrate equipment used in those determinations.
Specifications
Specifications
| Description | Chloride: <0,02mg/L Iodide: <0.01mg/L Nitrite: <0.01mg/L Nitrate: <0.01mg/L Phosphate: <0.02mg/L |
| Density | 1.00g/mL |
| Concentration | 1000 μg/mL of SO42− |
| IUPAC Name | disodium;sulfate |
| Quantity | 100 mL |
| Formula Weight | 142.04g/mol |
| Concentration or Composition (by Analyte or Components) | 1.479g Na2SO4 per 1L H2O |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction