Learn More
BUCHI Suction Module with Condensate Trap
Brand:  BUCHI 11055865
Product Code. 11967238
  • 959.00€ / Each
Estimated Shipment: 03-01-2025
to see stock.
Explore more special offers
Specifications
Suction Module
300 mL
For Aqueous Samples, Prevents Backflow of Condensate into the Sample Tube