missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Streptococcus pneumoniae Monoclonal Antibody (XI64)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA110833
This item is not returnable.
View return policy
Description
MA1-10833 detects Streptococcus pneumoniae. MA1-10833 has been successfully used in ELISA and Western blot applications. The MA1-10833 immunogen is Heat-killed S. pneumoniae strain R36A.
There are more than 85 antigenic types of S. pneumoniae, which are determined by capsule antigens. Altough S. pneumoniae is part of the normal respiratory flora, invasion commonly results in pneumonia as well as meningitis. S. pneumoniae is protected from phagocytosis by its polysaccharide capsule which is also a prominent virulence factor. Infections can usually be treated with antibiotics, although resistant strains are emerging.
Specifications
| Streptococcus pneumoniae | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Mouse | |
| Purified | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgM |
| ELISA | |
| XI64 | |
| Unconjugated | |
| Pneumococcus; s pneumoniae; S. pneumoniae | |
| Heat-killed S. pneumoniae strain R36A. | |
| 200 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction