missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Streptavidin, β-galactosidase conjugate

Streptavidin, β-galactosidase conjugate

Brand:  Molecular Probes™ S931

387.60 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.


Product Code. 10523283

  • 520.00€ / 1mg
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The streptavidin, ß-galactosidase conjugate can be used to detect biotin in a signal amplification scheme in conjunction with chromogenic substrates or fluorogenic substrates.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

beta-Galactosidase
Store in freezer (-5 to -30°C).
Solid
1 mg
Wet Ice
Streptavidin Conjugate (fluorescent)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.