Learn More
asecos™ Storage Box
Product Code. 17866446 Shop All asecos Products
Click to view available options
Depth (Metric):
475.00 mm
675.00 mm
875.00 mm
1080.00 mm
1280.00 mm
Unit Size:
Each
Product Code. 17866446
Brand: asecos™ 13101
This item requires a price update prior to being ordered.
Request a Price for this item here.
The Storage Box is for the storage of sealant mat.
  • Made of polyethylene
  • With handle and lid
  • Can be wall mounted
Specifications
Color Yellow
Material Polyethylene
Type Storage Box
Length (English) 7.48 in.
Length (Metric) 190.00 mm
Width (English) 7.48 in.
Width (Metric) 190.00 mm
Height (English) 8.27 in.
Height (Metric) 210.00 mm
Depth (English) 34.45 in.
Depth (Metric) 875.00 mm
Weight (English) 5.40 lb.
Weight (Metric) 2.45 kg