Learn More
Lenz Laborglasintrumente Stopcocks Straight Bore with PTFE Plug
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
63.00€ - 263.00€
Product Code | Brand | Plug Bore | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Plug Bore | Price | Quantity & Availability | |||||
10180531
|
Lenz Laborglasintrumente
02161402 |
2.5 mm |
263.00€
Pack of 10 |
Estimated Shipment: 10-01-2025
Log in to see stock. |
|||||
10313361
|
Lenz Laborglasintrumente
02162106 |
6 mm |
63.00€
Each |
Estimated Shipment: 20-12-2024
Log in to see stock. |
|||||
10222331
|
Lenz Laborglasintrumente
02162910 |
10 mm |
128.00€
Each |
Estimated Shipment: 10-01-2025
Log in to see stock. |
|||||
Description
- Acc. to DIN 12541-1
- With PTFE plug and screwthread retaining device
- One straight bore
- Material: borosilicate glass 3.3
- Leakage rate acc. to DIN 12540-1
Specifications
1-way stopcock | |
PTFE |