missing translation for 'onlineSavingsMsg'
Learn More

STF 31, Tocris Bioscience™

Product Code. 18095532
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

NAMPT inhibitor; also GLUT1 inhibitor

Inhibitor of GLUT1; inhibits glucose uptake in renal cell carcinoma (RCC) 4 cells. Activity causes necrotic cell death in von Hippel-Lindau (VHL)-deficient RCC cells. Also NAMPT inhibitor. Eliminates human pluripotent stem cells from culture with limited toxicity towards differentiated cells.

Chemical Identifiers

CAS 724741-75-7
Molecular Formula C23H25N3O3S
Molecular Weight (g/mol) 423.531
InChI Key NGQPRVWTFNBUHA-UHFFFAOYSA-N
PubChem CID 984333
IUPAC Name 4-[[(4-tert-butylphenyl)sulfonylamino]methyl]-N-pyridin-3-ylbenzamide
SMILES CC(C)(C)C1=CC=C(C=C1)S(=O)(=O)NCC2=CC=C(C=C2)C(=O)NC3=CN=CC=C3

Specifications

Quantity 50 mg
Synonym stf-31, stf 31, 4-4-1,1-dimethylethyl phenyl sulfonyl amino methyl-n-3-pyridinylbenzamide, stf31, stf hplc, 4-4-tert-butylphenylsulfonamido methyl-n-pyridin-3-yl benzamide, 4-4-tert-butylphenyl sulfonyl amino methyl-n-pyridin-3-yl benzamide, 4-4-tert-butylbenzenesulfonamido methyl-n-pyridin-3-yl benzamide, 4-4-tert-butylphenyl sulfonylamino methyl-n-pyridin-3-ylbenzamide, 2hj
Formula Weight 423.53
Percent Purity >99%
Chemical Name or Material STF 31
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'