Learn More
DWK Life Sciences STERIPLAN™ Petri Dish, from Soda-lime Glass
Shop All DWK Life Sciences Products
Click to view available options
Diameter (Metric):
40 mm
60 mm
80 mm
100 mm
120 mm
150 mm
180 mm
200 mm
Height (Metric):
10 mm
12 mm
15 mm
20 mm
25 mm
30 mm
45 mm
Unit Size:
Pack of 10
Description
Cultivation of microorganisms can take place on a gel-like nutrient medium, which is placed in a flat, round, transparent dish with lid (the so-called Petri dish). Due to the solid gel, mixing of the microorganisms is prevented and larger cell culture accumulations (colonies) can grow.
- Free from bubbles and streaks
- Even distribution of sewing bottom possible
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Material | Soda Lime Glass |
| For Use With (Application) | Growth and cultivation of microorganisms; Cultivation on solid gel culture medium; Microscopy of microorganisms; Cell culture |
| No. per Pack | 10/Pk. |
| Height (Metric) | 30 mm |
| Diameter (Metric) | 180 mm |