Learn More
Bel-Art™ Sterileware™ Liquid Thief
Shop All SP Scienceware Products
Click to view available options
Capacity (Metric):
100 mL
190 mL
Length (Metric):
50 cm
100 cm
Unit Size:
Pack of 20
Beschreibung
- Ideal for sampling water and other low viscosity liquids; 0.15 cm (0.0625 in.) tip hole diameter
- Made from FDA and EU conforming materials
- Safe for food, drug and cosmetic use
- High density polyethylene (HDPE) thief heat sealed in individual polyethylene bag
- Gamma irradiated sterile (SAL 10-6)
- Ready for one time use and disposal
- Lot stamped for accurate tracing
- Shipped with a Certificate of Processing for Sterilization
Spezifikation
Spezifikation
| Product Type | Liquid Sampler |
| Material | HDPE |
| Autoclavable | Not Autoclavable |
| Capacity (English) | 3.4 oz. |
| Capacity (Metric) | 100 mL |
| Sterility | Sterile |
| Quantity | 20/Pk. |
| Length (English) | 19.75 in. |
| Length (Metric) | 50 cm |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur