Learn More
Wheaton Science Products Steel Rack for 12 BOD Bottles 300ml
Brand:  Wheaton Science Products W227731
Steel Rack for 12 BOD Bottles 300ml
Product Code. 15507235
  • ONLINE EXCLUSIVE PRICE
    105.00€
    101.97€ / Each
    Save 3.03€
    2% Off
Only null left
Estimated Shipment: 17-02-2026